missing translation for 'onlineSavingsMsg'
Learn More
n-Decylamine, 99%, Thermo Scientific Chemicals Product Code.: 10606624

n-Decylamine, 99%, Thermo Scientific Chemicals

Product Code. 10606624
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10606624

Brand: Thermo Scientific Acros 111920500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-57-1
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00008149
InChI Key MHZGKXUYDGKKIU-UHFFFAOYSA-N
Synonym decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amineShow MoreShow Less
PubChem CID 8916
IUPAC Name decan-1-amine
SMILES CCCCCCCCCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 12°C to 15°C
Color Brown or Colorless to Yellow
Density 0.7870g/mL
Boiling Point 216°C to 218°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9NH2
Refractive Index 1.4350 to 1.437
Quantity 50 g
Beilstein 04, 199
Specific Gravity 0.787
Formula Weight 157.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Decylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466929 Certificate TypeCertificate of Analysis Date06/11/2024 Catalog Number111920500,111920100
Lot NumberA0466295 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number111920500,111920100
Lot NumberA0460006 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog Number111920500,111920100
Lot NumberA0453979 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number111920500,111920100
Lot NumberA0403019 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number111920500,111920100
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Decylamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.