missing translation for 'onlineSavingsMsg'
Learn More
n-Decyldimethylchlorosilane, 97%, Thermo Scientific™ Product Code.: 11395587

n-Decyldimethylchlorosilane, 97%, Thermo Scientific™

Product Code. 11395587
10 g
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11395587

Brand: Thermo Scientific Alfa Aesar 042769.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38051-57-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00054168
InChI Key ZLZGHBNDPINFKG-UHFFFAOYSA-N
Synonym chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysaShow MoreShow Less
PubChem CID 2758020
IUPAC Name chloro-decyl-dimethylsilane
SMILES CCCCCCCCCC[Si](C)(C)Cl

Specifications

Boiling Point 98°C (2mmHg)
Physical Form Liquid
Quantity 10 g
UN Number UN1760
Flash Point 137°C (279°F)
Formula Weight 234.88
Percent Purity 97%
Refractive Index 1.441
Sensitivity Moisture Sensitive
Density 0.866g/mL
Chemical Name or Material n-Decyldimethylchlorosilane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Decyldimethylchlorosilane, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.