Learn More
N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals
£20.60 - £62.41
Chemical Identifiers
CAS | 207291-76-7 |
---|---|
Molecular Formula | C18H19NO5 |
Molecular Weight (g/mol) | 329.35 |
MDL Number | MFCD00037139 |
InChI Key | GAPWKFLOMOFHGO-UHFFFAOYNA-N |
Synonym | n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate |
PubChem CID | 18528118 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate |
SMILES | O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474567
|
Thermo Scientific Alfa Aesar
B21107.06 |
5 g |
£20.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484567
|
Thermo Scientific Alfa Aesar
B21107.14 |
25 g |
£62.41
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Chemical Identifiers
207291-76-7 | |
329.35 | |
GAPWKFLOMOFHGO-UHFFFAOYNA-N | |
18528118 | |
O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
C18H19NO5 | |
MFCD00037139 | |
n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate |
Specifications
207291-76-7 | |
C18H19NO5 | |
2225975 | |
Insoluble in water. | |
−19° (c=1 in DMF) | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate | |
18528118 | |
98% |
∼130°C | |
MFCD00037139 | |
n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate | |
GAPWKFLOMOFHGO-UHFFFAOYNA-N | |
O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
329.35 | |
329.36 (311.34 Anhydrous) | |
N-Fmoc-L-alanine monohydrate |
Safety and Handling
missing translation for 'einecsNumber' : 252-660-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.