missing translation for 'onlineSavingsMsg'
Learn More
n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals Product Code.: 11373588

n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals

Product Code. 11373588
4 x 1 L
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11373588

Brand: Thermo Scientific Alfa Aesar 039199.M1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point °C to 95°C
Density 0.659 g/mL
Boiling Point 69°C
Quantity 4 x 1 L
Flash Point -23°C (-9°F)
Physical Form Liquid, Distilled In Glass
Refractive Index 1.375
Assay Percent Range 95% min
Beilstein 1730733
Solubility Information Very soluble in ethanol,ethyl ether and chloroform.
Formula Weight 86.18
Grade HPLC
Chemical Name or Material n-Hexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM30L709 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog Number039199.K7,039199.K2,039199.M1
Lot Number2404916 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number039199.K7,039199.K2,039199.M1
Lot Number2496240 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number039199.K7,039199.K2,039199.M1
Lot Number2402755 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number039199.K7,039199.K2,039199.M1
Lot Number2400124 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number039199.K7,039199.K2,039199.M1
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexane, HPLC Grade
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Hexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.