missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide, Thermo Scientific™
£8.15 - £13.00
Chemical Identifiers
CAS | 22179-86-8 |
---|---|
Molecular Formula | C8H7F3N2O |
Molecular Weight (g/mol) | 204.152 |
InChI Key | QCVFLUSIBKAKPC-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl benzamidoxime, n-hydroxy-4-trifluoromethyl-benzamidine, n'-hydroxy-4-trifluoromethyl benzenecarboximidamide, n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzene-1-carboximidamide, 4-trifluoromethylbenzamidoxime, hydroxyimino 4-trifluoromethyl phenyl methylamine, 4-trifluoromethylbenzamideoxime, benzenecarboximidamide,n-hydroxy-4-trifluoromethyl |
PubChem CID | 9581576 |
IUPAC Name | N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide |
SMILES | C1=CC(=CC=C1C(=NO)N)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10147352
|
Thermo Scientific Maybridge
RF03217DA |
1 g |
£8.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10733557
|
Thermo Scientific Maybridge
RF03217DE |
5 g | |||||||
Description
Chemical Identifiers
22179-86-8 | |
204.152 | |
4-trifluoromethyl benzamidoxime, n-hydroxy-4-trifluoromethyl-benzamidine, n'-hydroxy-4-trifluoromethyl benzenecarboximidamide, n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzene-1-carboximidamide, 4-trifluoromethylbenzamidoxime, hydroxyimino 4-trifluoromethyl phenyl methylamine, 4-trifluoromethylbenzamideoxime, benzenecarboximidamide,n-hydroxy-4-trifluoromethyl | |
N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide |
C8H7F3N2O | |
QCVFLUSIBKAKPC-UHFFFAOYSA-N | |
9581576 | |
C1=CC(=CC=C1C(=NO)N)C(F)(F)F |
Specifications
22179-86-8 | |
C8H7F3N2O | |
QCVFLUSIBKAKPC-UHFFFAOYSA-N | |
N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide | |
9581576 | |
Tech. | |
N'-Hydroxy-4-(trifluoromethyl)benzenecarboximidamide |
90% | |
4-trifluoromethyl benzamidoxime, n-hydroxy-4-trifluoromethyl-benzamidine, n'-hydroxy-4-trifluoromethyl benzenecarboximidamide, n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzimidamide, z-n'-hydroxy-4-trifluoromethyl benzene-1-carboximidamide, 4-trifluoromethylbenzamidoxime, hydroxyimino 4-trifluoromethyl phenyl methylamine, 4-trifluoromethylbenzamideoxime, benzenecarboximidamide,n-hydroxy-4-trifluoromethyl | |
C1=CC(=CC=C1C(=NO)N)C(F)(F)F | |
204.152 | |
204.15 | |
Technical |
Safety and Handling
GHS Signal Word: Warning