missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™ Product Code.: 11396805

N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 11396805
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11396805

Brand: Thermo Scientific Maybridge CC58146CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-54-3
Molecular Formula C8H10BrNO2S
Molecular Weight (g/mol) 264.137
MDL Number MFCD08690308
InChI Key FJHBRYOEMIQSPC-UHFFFAOYSA-N
Synonym n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamineShow MoreShow Less
PubChem CID 18525892
IUPAC Name 1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
SMILES CNCC1=C2C(=C(S1)Br)OCCO2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 264.14
Percent Purity 90%
Chemical Name or Material N-methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.