missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals Product Code.: 11434514

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals

Product Code. 11434514
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11434514

Brand: Thermo Scientific Alfa Aesar A13141.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfmShow MoreShow Less
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Density 1.076
Boiling Point 130°C to 132°C
Flash Point 25°C (77°F)
Refractive Index 1.38
Quantity 5 g
UN Number UN1993
Beilstein 1941550
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253729 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberA13141.06,A13141.22,A13141.14
Lot Number10249265 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberA13141.06,A13141.22,A13141.14
Lot Number10231513 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog NumberA13141.06,A13141.22,A13141.14
Lot Number10245564 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA13141.06,A13141.22,A13141.14
Lot Number10236212 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog NumberA13141.06,A13141.22,A13141.14
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)-Acetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.