Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals
£64.00 - £581.00
Chemical Identifiers
CAS | 24589-78-4 |
---|---|
Molecular Formula | C6H12F3NOSi |
Molecular Weight (g/mol) | 199.25 |
MDL Number | MFCD00000411 |
InChI Key | MSPCIZMDDUQPGJ-UHFFFAOYSA-N |
Synonym | mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm |
PubChem CID | 32510 |
ChEBI | CHEBI:85064 |
IUPAC Name | 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide |
SMILES | CN(C(=O)C(F)(F)F)[Si](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10226850
|
Thermo Scientific Acros
221580050 |
5 g | Glass bottle |
£64.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236850
|
Thermo Scientific Acros
221580250 |
25 g | Glass Bottle |
£204.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781394
|
Thermo Scientific
221581000 |
100 g | Glass bottle |
£581.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, Silylation reagentChemical Identifiers
24589-78-4 | |
199.25 | |
MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
32510 | |
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide |
C6H12F3NOSi | |
MFCD00000411 | |
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
CHEBI:85064 | |
CN(C(=O)C(F)(F)F)[Si](C)(C)C |
Specifications
24589-78-4 | |
100.0 | |
1.0700g/mL | |
25°C | |
96% min. (GC) | |
CF3CON(CH3)Si(CH3)3 | |
MFCD00000411 | |
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide | |
32510 | |
199.25 | |
Liquid |
96.0 | |
Colorless to Yellow | |
130°C to 132°C | |
Authentic | |
C6H12F3NOSi | |
1.3782 to 1.3802 | |
1.07 | |
Solubility in water: reacts | |
CN(C(=O)C(F)(F)F)[Si](C)(C)C | |
199.25 | |
CHEBI:85064 | |
97% | |
N-Methyl-N-(trimethylsilyl)trifluoroacetamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-331-6
- N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only