Learn More
N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 184.0°C to 187.0°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 250 g |
Formula Weight | 262.33 |
Percent Purity | 99% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463139 | Certificate of Analysis | 23/05/2024 | 395002500,395000500,395000010 |
A0461273 | Certificate of Analysis | 19/03/2024 | 395002500,395000500,395000010 |
A0459668 | Certificate of Analysis | 26/01/2024 | 395002500,395000500,395000010 |
A0456691 | Certificate of Analysis | 09/10/2023 | 395002500,395000500,395000010 |
A0455838 | Certificate of Analysis | 25/08/2023 | 395002500,395000500,395000010 |
Safety and Handling

- N,N-Diethyl-p-phenylenediamine sulfate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.