missing translation for 'onlineSavingsMsg'
Learn More
N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals Product Code.: 10197133

N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals

Product Code. 10197133
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10197133

Brand: Thermo Scientific Acros 395000500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H16N2·H2SO4
Molecular Weight (g/mol) 262.33
MDL Number MFCD00012993
InChI Key AYLDJQABCMPYEN-UHFFFAOYSA-N
Synonym n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1Show MoreShow Less
PubChem CID 80166
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid
SMILES CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Formula Weight 262.33
Percent Purity 99%
Physical Form Crystalline Powder or Crystals With Lumps
Chemical Name or Material N, N-Diethyl-p-phenylenediamine sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463139 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number395002500,395000500,395000010
Lot NumberA0461273 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number395002500,395000500,395000010
Lot NumberA0459668 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog Number395002500,395000500,395000010
Lot NumberA0456691 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number395002500,395000500,395000010
Lot NumberA0455838 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number395002500,395000500,395000010
compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.