Learn More
N,N-Diethylaniline, 99%, Thermo Scientific Chemicals
£31.36 - £275.37
Chemical Identifiers
CAS | 91-66-7 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.24 |
MDL Number | MFCD00009042,MFCD31699978 |
InChI Key | GGSUCNLOZRCGPQ-UHFFFAOYSA-N |
Synonym | diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine |
PubChem CID | 7061 |
IUPAC Name | N,N-diethylaniline |
SMILES | CCN(CC)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413204
|
Thermo Scientific Alfa Aesar
A10560.AP |
500 mL |
£31.36
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423204
|
Thermo Scientific Alfa Aesar
A10560.0F |
2500 mL |
£88.24
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433204
|
Thermo Scientific Alfa Aesar
A10560.0D |
10,000 mL |
£275.37
10000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.
Notes
Incompatible with strong oxidizing agents and strong acids. .
Chemical Identifiers
91-66-7 | |
149.24 | |
GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
7061 | |
CCN(CC)C1=CC=CC=C1 |
C10H15N | |
MFCD00009042,MFCD31699978 | |
diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
N,N-diethylaniline |
Specifications
91-66-7 | |
0.936 | |
85°C (185°F) | |
1.542 | |
UN2432 | |
14,3114 | |
Miscible with water and acetone. Slightly miscible with chloroform,alcohol and ether. | |
CCN(CC)C1=CC=CC=C1 | |
149.24 | |
149.24 | |
N,N-Diethylaniline |
-38°C | |
215°C to 216°C | |
C10H15N | |
MFCD00009042,MFCD31699978 | |
742483 | |
diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
N,N-diethylaniline | |
7061 | |
99% |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N,N DIETHYLANILINE
missing translation for 'einecsNumber' : 202-088-8
missing translation for 'rtecsNumber' : BX3400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.