Learn More
N,N-Diisopropylethylamine, 99+%, Thermo Scientific Chemicals
£15.40 - £1636.00
Chemical Identifiers
CAS | 7087-68-5 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.24 |
MDL Number | MFCD00008868 |
InChI Key | JGFZNNIVVJXRND-UHFFFAOYSA-N |
Synonym | n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine |
PubChem CID | 81531 |
IUPAC Name | N-ethyl-N-propan-2-ylpropan-2-amine |
SMILES | CCN(C(C)C)C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10142860
|
Thermo Scientific Acros
115220250 |
25 g | Glass bottle |
£15.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10734991
|
Thermo Scientific Acros
115221000 |
100 g | Glass bottle |
£41.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10244620
|
Thermo Scientific Acros
115225000 |
500 g | Glass bottle |
£124.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10008129
|
Thermo Scientific Acros
115220026 |
2.5 kg | Plastic Bottle |
£448.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12664947
|
Thermo Scientific Acros
115220101 |
10 kg | Plastic Can |
£1636.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7087-68-5 | |
129.24 | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
81531 | |
CCN(C(C)C)C(C)C |
C8H19N | |
MFCD00008868 | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
N-ethyl-N-propan-2-ylpropan-2-amine |
Specifications
7087-68-5 | |
100.0 | |
Colorless to Yellow | |
127.0°C | |
Authentic | |
C8H19N | |
1.4123 to 1.4143 | |
04,IV,511 | |
15,3219 | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
N-ethyl-N-propan-2-ylpropan-2-amine | |
81531 | |
129.24 | |
Liquid |
98.0 | |
-46.0°C | |
0.7560g/mL | |
6°C | |
99% min. (GC) | |
[(CH3)2CH]2NC2H5 | |
MFCD00008868 | |
01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176 | |
0.756 | |
Solubility in water: 3.9g/L (20°C). | |
CCN(C(C)C)C(C)C | |
129.24 | |
0.88 mm2/s (20°C) | |
99+% | |
N,N-Diisopropylethylamine, 98+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 230-392-0
missing translation for 'tsca' : TSCA
- N,N-Diisopropylethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only