missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

£53.97 - £779.88

Chemical Identifiers

CAS 536-46-9
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
MDL Number MFCD00012991
InChI Key PXJHVKRLFWZUNV-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PubChem CID 2724166
SMILES [H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444133
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10343.14
25 g
£53.97
25g
Please to purchase this item. Need a web account? Register with us today!
11454133
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10343.22
100 g
£189.95
100g
Please to purchase this item. Need a web account? Register with us today!
11464133
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10343.36
500 g
£779.88
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

Solubility
Soluble in water

Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
Specifications

Chemical Identifiers

536-46-9
209.11
PXJHVKRLFWZUNV-UHFFFAOYSA-N
2724166
C8H14Cl2N2
MFCD00012991
n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
[H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1

Specifications

536-46-9
C8H14Cl2N2
UN2811
n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PXJHVKRLFWZUNV-UHFFFAOYSA-N
4-N,4-N-dimethylbenzene-1,4-diamine;dihydrochloride
2724166
98%
212°C (decomposition)
MFCD00012991
Hygroscopic; Light sensitive
Soluble in water
[H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
209.11
209.12
N,N-Dimethyl-p-phenylenediamine dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P361-P363-P501c

H300+H330-H311-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 208-635-7

missing translation for 'rtecsNumber' : ST1300000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • N,N-Dimethyl-p-phenylenediamine dihydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethyl-p-phenylenediamine dihydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only