missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals Product Code.: 12337083

N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals

Product Code. 12337083
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 12337083

Brand: Thermo Scientific Acros 448381000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -61.0°C
CAS 68-12-2
Density 0.9450g/mL
Boiling Point 153.0°C
Quantity 100 mL
Flash Point 58°C
Identification Pass Test
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4290 to 1.4320
Assay Percent Range 99.8%
Residue after Evaporation 0.001 % max.
Specific Gravity 0.945
Molecular Formula C3H7NO
Linear Formula HCON(CH3)2
MDL Number MFCD00003284
Viscosity 0.8 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.10
PubChem CID 6228
ChEBI CHEBI:17741
Formula Weight 73.09
Percent Purity 99.8%
Grade Extra Dry
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2524260 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number448381000
Lot Number2412083 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number448381000
Lot Number2406182 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number448381000
Lot Number2402023 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number448381000
Lot Number2415080 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number448381000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Dimethylformamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.