missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide, ACS, 99.8+%, Thermo Scientific Chemicals Product Code.: 11333498

N,N-Dimethylformamide, ACS, 99.8+%, Thermo Scientific Chemicals

Product Code. 11333498
4 x 4 L
Click to view available options
Quantity:
4 L
4 x 4 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11333498

Brand: Thermo Scientific Alfa Aesar 039117.M6

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

A solvent for chromogenic substrates

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfaShow MoreShow Less
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7
Melting Point -61°C
Density 0.944
Boiling Point 153°C
Quantity 4 x 4 L
Flash Point 57°C (135°F)
Odor Amine-like
Refractive Index 1.431
UN Number UN2265
Beilstein 605365
Merck Index 14,3243
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L031 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number039117.AP,039117.K7,039117.K2,039117.M6,039117.M1
Lot NumberZ31K007 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number039117.AP,039117.K7,039117.K2,039117.M6,039117.M1
Lot NumberX16K031 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number039117.AP,039117.K7,039117.K2,039117.M6,039117.M1
Lot NumberW03K007 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number039117.AP,039117.K7,039117.K2,039117.M6,039117.M1
Lot NumberT18K003 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number039117.AP,039117.K7,039117.K2,039117.M6,039117.M1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide, ACS, 99.8+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.