missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS, 99.8+%, Thermo Scientific Chemicals

A solvent For Chromogenic Substrates

£30.75 - £150.75

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490211
View Documents
Thermo Scientific Alfa Aesar
039117.AP
500 mL
£30.75
500mL
Please to purchase this item. Need a web account? Register with us today!
11313498
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039117.K2
1 L
£50.45
1 L
Please to purchase this item. Need a web account? Register with us today!
11323498
View Documents
Thermo Scientific Alfa Aesar
039117.M1
4 x 1 L
£150.75
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities
  • Appearance Clear
  • Color (APHA) 15
  • Evaporation residue 0.005%
  • Titratable base 0.003meq/g
  • Titratable acid 0.0005meq/g
  • H2O 0.15%
Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O

Specifications

68-12-2
Colorless
153°C
440°C (824°F)
Amine-like
1.431
605365
Miscible with water and most organic solvents.
CN(C)C=O
73.10
CHEBI:17741
3.5 hPa (3mm Hg) at 20°C
Liquid
°C to 61°C
0.944 g/cm3 at 20°C
57°C (135°F)
>99.8%
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09
ACS
N, N-Dimethylformamide, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c

H226-H312+H332-H319-H335-H336-H350-H360D

missing translation for 'dotInformation' : DOT Class: 3, Packaging Group: III

missing translation for 'einecsNumber' : 200-679-5

missing translation for 'rtecsNumber' : LQ2100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • N,N-Dimethylformamide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Dimethylformamide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only