missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylbenzylamine, 99%, Thermo Scientific Chemicals
£18.79 - £493.22
Chemical Identifiers
CAS | 103-32-2 |
---|---|
Molecular Formula | C13H13N |
Molecular Weight (g/mol) | 183.25 |
MDL Number | MFCD00003018 |
InChI Key | GTWJETSWSUWSEJ-UHFFFAOYSA-N |
Synonym | n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl anilineShow More |
PubChem CID | 66028 |
IUPAC Name | N-benzylaniline |
SMILES | C1=CC=C(C=C1)CNC2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10457022
|
Thermo Scientific Acros
417390050 |
5 g | Glass bottle |
£18.79
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10528332
|
Thermo Scientific Acros
417390250 |
25 g | Glass bottle |
£33.37
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10657401
|
Thermo Scientific Acros
417391000 |
100 g | Glass bottle |
£121.61
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10679004
|
Thermo Scientific Acros
417395000 |
500 g | Glass Bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-32-2 | |
183.25 | |
GTWJETSWSUWSEJ-UHFFFAOYSA-N | |
66028 | |
C1=CC=C(C=C1)CNC2=CC=CC=C2 |
C13H13N | |
MFCD00003018 | |
n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl anilineShow More | |
N-benzylaniline |
Specifications
103-32-2 | |
100.0 | |
Beige-Yellow to Colorless | |
306°C to 307°C | |
Authentic | |
C13H13N | |
MFCD00003018 | |
15, 1129 | |
n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline | |
GTWJETSWSUWSEJ-UHFFFAOYSA-N | |
N-benzylaniline | |
66028 | |
99% | |
N-Phenylbenzylamine |
98.5 | |
34°C to 38°C | |
1.0600g/mL | |
152°C | |
98.5% min. (GC) | |
C6H5CH2NHC6H5 | |
12, 1023 | |
1.06 | |
Solubility in water: insoluble in water. Other solubilities: very soluble in ethanol,ether,soluble in chloroform | |
C1=CC=C(C=C1)CNC2=CC=CC=C2 | |
183.25 | |
183.25 | |
Crystalline Powder or Low Melting Mass |
Safety and Handling
missing translation for 'einecsNumber' : 203-100-4
Product Identifier
- N-Phenylbenzylamine
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Phenylbenzylamine, 99%, Thermo Scientific Chemicals