Learn More
N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals
£45.93 - £178.89
Chemical Identifiers
CAS | 1664-40-0 |
---|---|
Molecular Formula | C8H12N2 |
Molecular Weight (g/mol) | 136.2 |
MDL Number | MFCD00008162 |
InChI Key | OCIDXARMXNJACB-UHFFFAOYSA-N |
Synonym | n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamine |
PubChem CID | 74270 |
IUPAC Name | N'-phenylethane-1,2-diamine |
SMILES | C1=CC=C(C=C1)NCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173560
|
Thermo Scientific Acros
130610100 |
10 mL | Glass bottle |
£45.93
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10147893
|
Thermo Scientific Acros
130610500 |
50 mL | Glass bottle |
£178.89
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1664-40-0 | |
136.2 | |
OCIDXARMXNJACB-UHFFFAOYSA-N | |
74270 | |
C1=CC=C(C=C1)NCCN |
C8H12N2 | |
MFCD00008162 | |
n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamine | |
N'-phenylethane-1,2-diamine |
Specifications
1664-40-0 | |
100.0 | |
1.0400g/mL | |
>112°C | |
99% | |
C6H5NHCH2CH2NH2 | |
MFCD00008162 | |
1.04 | |
OCIDXARMXNJACB-UHFFFAOYSA-N | |
N'-phenylethane-1,2-diamine | |
74270 | |
99% | |
N-Phenylethylenediamine |
98.5 | |
Yellow | |
262.0°C to 264.0°C | |
Authentic | |
C8H12N2 | |
1.5855 to 1.5875 | |
12, 543 | |
n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamine | |
C1=CC=C(C=C1)NCCN | |
136.2 | |
136.2 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-773-4
- N-Phenylethylenediamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only