missing translation for 'onlineSavingsMsg'
Learn More
N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals Product Code.: 10173560

N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 10173560
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10173560

Brand: Thermo Scientific Acros 130610100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1664-40-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00008162
InChI Key OCIDXARMXNJACB-UHFFFAOYSA-N
Synonym n-phenylethylenediamine, n1-phenylethane-1,2-diamine, n-2-aminoethyl aniline, 1,2-ethanediamine, n-phenyl, ethylenediamine, n-phenyl, n-phenyl-1,2-ethanediamine, benzenamine, n-2-aminoethyl, n-phenylethane-1,2-diamine, 1,2-ethanediamine, n1-phenyl, 2-aminoethyl phenylamineShow MoreShow Less
PubChem CID 74270
IUPAC Name N'-phenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)NCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0400g/mL
Boiling Point 262.0°C to 264.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5NHCH2CH2NH2
Refractive Index 1.5855 to 1.5875
Quantity 10 mL
Beilstein 12, 543
Specific Gravity 1.04
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Phenylethylenediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0447956 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number130610500,130610100
Lot NumberA0447956 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog Number130610500,130610100
Lot NumberA0455796 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number130610500,130610100
Lot NumberA0429204 Certificate TypeCertificate of Analysis Date19/04/2022 Catalog Number130610500,130610100
Lot NumberA0415742 Certificate TypeCertificate of Analysis Date15/01/2021 Catalog Number130610500,130610100
compliance-icons
Product Identifier
  • N-Phenylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.