Learn More
NAD+, MedChemExpress
MedChemExpress NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.
Brand:  Medchem Express HY-B0445
Encompass_Preferred
 View more versions of this product
Product Code. 16400938
  • £205.70 / 5g
Explore more special offers
Description
Description
NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.
Specifications
Specifications
NAD+
White
C21H27N7O14P2
β-DPN β-NAD β-Nicotinamide Adenine Dinucleotide
O[C@H]([C@@H]1O)[C@@H](O[C@@H]1COP(OP(OC[C@H]2O[C@@H]([N+]3=CC=CC(C(N)=O)=C3)[C@H](O)[C@@H]2O)([O-])=O)(O)=O)N4C(N=CN=C5N)=C5N=C4
663.43
Research
Metabolism-protein/nucleotide metabolism
53-84-9
Research
5 g
H2O : 41.67 mg/mL (62.81 mM; Need ultrasonic)
663.43
98.24%
Powder