Learn More
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
GREENER_CHOICE
View contents easily with these autoclavable polypropylene desiccators, featuring a transparent PC cover and robust body resistant to all solid desiccants.
£96.62 - £130.00
Specifications
Autoclavable Autoclavable
Cabinet Style Top Loading
Material Polypropylene, Polycarbonate
No. per Pack 1
View More Specs
Alert:
Do not use concentrated acids as desiccants.
Products 2
Product Code Brand Height (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Height (Metric) Diameter (Metric) Inner Price Quantity & Availability  
10451751
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
5309-0250
262 mm 246 mm
£96.62
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
10548842
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
5315-0150
149 mm 149 mm
£130.00
Each
In Stock
to see stock.
Add to basket
Description
Description
View contents easily with Thermo Scientific™ Autoclavable Polypropylene Desiccators, featuring a robust body and high resistance to all solid desiccants. These desiccators have a transparent polycarbonate cover. The small size (5315-0150) comes with a 140 mm aluminum plate. The plate for the large size (5309-0250) is sold separately.
  • Clear polycarbonate (PC) cover has recessed rim to keep grease off benchtop when cover is set down
  • No gasket required, only grease is needed
  • Large desiccator (Cat. No. 5310-0250) takes any standard 230mm desiccator plate such as Cat. No. 5312-0230 (sold separately)
  • Autoclavable; repeated autoclaving will reduce the life of the product
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Specifications
Autoclavable
Top Loading
Polypropylene, Polycarbonate
1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover