missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Biohazardous Waste Containers
GREENER_CHOICE

Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.

Brand:  Thermo Scientific™ 6370-0015

223.54 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10481022

  • £532.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).

Features:

  • Will not rust, dent or puncture like other materials
  • Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
  • Autoclavable/Biohazard
  • Symbol printed in English and Spanish

.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 gal.
13.25 in.
Polypropylene
Waste Container Biohazardous
74 cm
1
U.S. OSHA Standard 29 CFR Part 1910.1030
57 L
Non-sterile
29 in.
1
34 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.