missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Biohazardous Waste Containers
GREENER_CHOICE

Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.

Brand:  Thermo Scientific™ 6370-0004

143.63 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.


 View more versions of this product

Product Code. 10556551

  • £263.00 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).

Features:

  • Will not rust, dent or puncture like other materials
  • Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
  • Autoclavable/Biohazard
  • Symbol printed in English and Spanish

.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 gal.
8.75 in.
Polypropylene
Waste Container Biohazardous
28 cm
1
U.S. OSHA Standard 29 CFR Part 1910.1030
5.5 L
Non-sterile
11 in.
1
22 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers