Learn More
Thermo Scientific™ Nalgene™ Biohazardous Waste Containers
Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.
Brand: Thermo Scientific™ 6370-0005
211.20 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 54
Additional Details : Weight : 2.42100kg
Description
Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).
Features:
- Will not rust, dent or puncture like other materials
- Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
- Autoclavable/Biohazard
- Symbol printed in English and Spanish
.
Specifications
5 gal. | |
Waste Container Biohazardous PP 5 gallon | |
19 L | |
Non-sterile | |
17 in. | |
1 | |
28 cm |
U.S. OSHA Standard 29 CFR Part 1910.1030 | |
11 in. | |
Polypropylene | |
Waste Container Biohazardous | |
43 cm | |
1 |