Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners
Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Brand: Thermo Scientific™ 343050-0080
703.26 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.
- Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
- Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic

Specifications
Tank Liner | |
80 gal. | |
Nalgene cylindrical tanks from 5 gal (19L) to 200 | |
10 |
Polyethylene | |
303 L | |
10 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7438619010 | Certificate of Processing | 22/01/2025 | 343050-0080 |
Special Offers
-
Multiple Manufacturers
Outlet Corner
Use code 24095 to receive this promotion.Promo Code: 24095Expires: 2025-04-15
Days
9Hours
21Minutes
55Seconds
4
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.