Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Brand: Thermo Scientific™ CS509X4
562.08 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
- Warranty varies by country, please contact us for details

Specifications
Cryogenic Storage Box | |
2 mL | |
2 in. | |
2 mL Vials |
7.06 cu. ft. | |
Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus | |
81 | |
40/Cs. |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.