Learn More
Thermo Scientific™ CryoTube™ Holders
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
Brand: Thermo Scientific™ 5030-0505
25.02 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
- Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
- Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
Yes | |
Polycarbonate | |
Cryo Tube | |
Nalgene™ | |
5 x 5 |
Blue | |
Vial Holder | |
25 Cryogenic Tubes | |
1/Pk., 4/Cs. |