Learn More
Thermo Scientific™ Nalgene™ HDPE, Jerry Can with Tethered Polypropylene Closure (Double Port)
For Use With: Ideal as a reservoir in automated systems; Closure Material: Polypropylene; Color: White
Brand: Thermo Scientific™ 2243-9013
Description
Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-ported 2243-9013 is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38mm closure is used for refilling or emptying the reservoir.
Key Features:- Jerrican is ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
- Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing
- Molded-in handle provides a sturdy grip when carrying heavy contents
- Integral shoulder handle and molded bottom grip make lifting comfortable
- Wide stance and low center of gravity add stability
- Heavy rugged design with good chemical resistance allows for a wide variety of solutions
- Space-saving rectangular shape permits efficient storage
Ordering Information:
- For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
- Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
- Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator

Specifications
3.5 gal. | |
USP Class VI Compliant/FDA21 CFR 177.1520 Compliant | |
Polypropylene | |
Tethered | |
Jerrican w/double port | |
38 mm | |
HDPE | |
1 Ea. |
13 L | |
White | |
53B | |
White | |
1.5 in. | |
Ideal as a reservoir in automated systems | |
4/Cs. | |
Jerry Can |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1422854 | Certificate of Compliance | 22/02/2025 | 2243-9013 |
1310761 | Certificate of Compliance | 10/12/2024 | 2243-9013 |
1323447 | Certificate of Compliance | 10/12/2024 | 2243-9013 |
1407863 | Certificate of Compliance | 30/07/2024 | 2243-9013 |
1401161 | Certificate of Compliance | 12/02/2024 | 2243-9013 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.