missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Low-Profile Carboy
GREENER_CHOICE

For Use With: Dispense most liquids, including reagents, distilled water and titrants.; Closure Material: Polypropylene; Color: White

£147.96 - £577.00

Specifications

Autoclavable Not Autoclavable
Closure Material Polypropylene
Color White
For Use With (Application) Dispense most liquids, including reagents, distilled water and titrants.
Material HDPE
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10170341
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2323-0015
15 L N/A N/A N/A
10231901
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2323-0008
8 L N/A N/A N/A
15941491
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2323-0008
8 L N/A N/A N/A
Description

Description

Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with most aqueous and alcohol lab liquids, including reagents, distilled water and titrants. Convenient integral carry handle for safe, easy transport. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:
  • Stack up to three high to save space on benchtop or in refrigerator
  • Contour handle allows easy one-handed carrying—even when full
  • Excellent chemical resistance—can tolerate strong acids and bases for a wide variety of solutions
  • Leakproof† when used with Nalgene closures
  • Carboy and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Spigot and spigot closure and four acid-proof, pressure-sensitive labels

Note:

Nalgene rugged HDPE carboys are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE carboys and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE carboys.

Specifications

Specifications

Not Autoclavable
White
HDPE
Polypropylene
Dispense most liquids, including reagents, distilled water and titrants.
Low-profile Carboy
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ HDPE, Low-Profile Carboy

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.