missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
GREENER_CHOICE

Eliminate the need for a rubber stopper while maintaining a leakproof seal with these lined, HDPE serum vial closures, for use with continuous thread PETG serum vials.

Brand:  Thermo Scientific™ 312158-0022

1062.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10262333

  • £1420.82 / Pack of 2000
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Eliminate the need for a rubber stopper, yet maintain a leakproof† seal with Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closures for Continuous-Thread Vials. These nonsterile, high density polyethylene closures are for use with continuous-thread PETG serum vials.

Features:

  • Three-ply co-extruded liner (LDPE core between two facings of solid HDPE)
  • For use with nonsterile Nalgene PETG serum vials

Ordering Information:

Serum vial closures are bulk packed 2,000 per case.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
22.1 mm
Non-sterile
2000
HDPE
14.2 mm
ISO 13485:2003
20-415
Serum vial closures
Black
HDPE
2000
Nalgene
Screw Cap
2000 Cs.
Serum Vial
Nonsterile Nalgene PETG Serum Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile > Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.