Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Economy Bottles

Description
Contain samples and daily working solutions like buffers and bench reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Economy Bottles, lightweight versions of the original Nalgene Lab Quality Bottle. Suitable for lighter-duty, general purpose applications, these high-density polyethylene bottles are leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
- Linerless polypropylene caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
- Recommended for sample collection and storage of daily working solutions
- Bottles participate in My Green Labs' ACT Label program for Accountability, Consistency and Transparency around product sustainability
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 2 oz., 60 mL |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Color | Translucent |
Description | Narrow-Mouth Economy Bottle |
Diameter (Metric) Inner Neck | 14 mm |
Material | HDPE |
No. per Case | 72 |
No. per Pack | 72 |
Sterility | Non-sterile |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1423573 | Certificate of Compliance | 13/03/2025 | 2089-0002 |
1422550 | Certificate of Compliance | 04/02/2025 | 2089-0002 |
1412564 | Certificate of Compliance | 30/08/2024 | 2089-0002 |
1407807 | Certificate of Compliance | 13/06/2024 | 2089-0002 |
1396707 | Certificate of Compliance | 28/12/2023 | 2089-0002 |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.