Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Store, ship and package liquids with these highly reliable, lab-quality bottles, specifically designed for long-term demanding lab use.
Brand: Thermo Scientific™ 2002-0006
17.79 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 55
Additional Details : Weight : 0.30600kg
Description
Store, ship, and package liquids using highly reliable and durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
- Versatile and indispensable in the lab
- Uniform walls provide durability and resistance to splitting or punctures
- Can be used in a freezer up to -100°C (-149°F)
- Linerless polypropylene closures provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
- Participates in the ACT Label Program
- Leakproof†
Specifications
6 oz., 175 mL | |
6 oz. | |
Lab Quality Narrow-Mouth Bottle | |
HDPE | |
12 | |
Natural | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Narrow | |
Yes | |
Narrow Mouth Lab Quality Bottle | |
Unlined | |
24 mm | |
Screw Cap |
175 mL | |
Translucent | |
18 mm | |
72 | |
Non-sterile | |
24 mm | |
Yes | |
Round | |
125 mm | |
4.92 in. | |
Polypropylene | |
54 mm | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.