missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle Product Code.: 16450601
Estimated Shipment: 21-08-2025
to see stock.
Add to basket

Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle

Product Code. 16450601
5 mL
Click to view available options
Capacity (Metric):
5 mL
20 mL
This item is not returnable. View return policy

Product Code. 16450601

Brand: Thermo Scientific™ 38350005

Estimated Shipment: 21-08-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle for storage of high-valued pharmaceutical samples

  • Perfect for QA lot sample storage of high-valued pharmaceuticals
  • Small-volume bottles are molded of the same materials as larger containers, but require a lower volume of expensive reagents to perform validation testing
  • Validated to USP 788
  • Impact Resistant
  • < 5 EU/mL Endotoxin
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Color Clear
For Use With (Application) Fluid Storage and Transport
Material Polycarbonate
No. per Case 416
No. per Pack 52
Quantity 416/Case
Sterility Sterile
Weight (English) 9 lb.
Certifications/Compliance USP 788
Autoclavable Not Autoclavable
Product Type Validation Bottle
Width (English) 12.5 in.
Length (English) 12.5 in.
Height (English) 11 in.
Closure Size 20 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.