missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle
Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle for storage of high-valued pharmaceutical samples
£2535.00 - £5510.00
Specifications
Color | Clear |
---|---|
For Use With (Application) | Fluid Storage and Transport |
Material | Polycarbonate |
Sterility | Sterile |
Certifications/Compliance | USP 788 |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
16450601
|
Thermo Scientific™
383500-05 |
5 mL |
£5510.00
Pack of 416 |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
16460601
|
Thermo Scientific™
383500-20 |
20 mL |
£2535.00
Pack of 504 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
Description
- Perfect for QA lot sample storage of high-valued pharmaceuticals
- Small-volume bottles are molded of the same materials as larger containers, but require a lower volume of expensive reagents to perform validation testing
- Validated to USP 788
- Impact Resistant
- < 5 EU/mL Endotoxin
Specifications
Clear | |
Polycarbonate | |
USP 788 | |
Validation Bottle |
Fluid Storage and Transport | |
Sterile | |
Not Autoclavable | |
20 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle