Learn More
Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.
£22.65 - £777.00
Specifications
Color | Clear |
---|---|
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Material | PETG |
Sterility | Sterile |
Temperature Range (Metric) | -70°C to 70°C |
Product Code | Brand | Product Type | Capacity (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Capacity (Metric) | Quantity | Price | Quantity & Availability | |||
11319694
|
Thermo Scientific™
3025-42 |
Square Bottle | 125 mL | 20 x 5 Bottles | N/A | ||||
10791310
|
Thermo Scientific™
3005-42 |
Square Bottle | 500 mL | 14 x 5 Bottles |
£442.00
Pack of 70 |
In Stock
Log in to see stock. |
|||
10237747
|
Thermo Scientific™
3005-70 |
Square Bottle | 500 mL | 70 Bottles |
£550.00
Pack of 70 |
In Stock
Log in to see stock. |
|||
12059987
|
Thermo Scientific™
3110-42 |
Square Bottle | 1 L | 7 x 5 Bottles |
£777.00
Pack of 35 |
In Stock
Log in to see stock. |
|||
10329587
|
Thermo Scientific™
3110-35 |
Square Bottle | 1 L | 35 Bottles |
£491.00
Pack of 35 |
In Stock
Log in to see stock. |
|||
10524408
|
Thermo Scientific™
3230-42 |
Square Bottle | 2 L | 4 x 5 Bottles |
£368.00
Pack of 20 |
In Stock
Log in to see stock. |
|||
10297887
|
Thermo Scientific™
3230-20 |
Square Bottle | 2 L | 20 Bottles |
£356.00
Pack of 20 |
In Stock
Log in to see stock. |
|||
10440568
|
Thermo Scientific™
3415-42 |
Square Bottle | 5 L | 6 Bottles |
£426.00
Pack of 6 |
In Stock
Log in to see stock. |
|||
10553298
|
Thermo Scientific™
3415-16 |
Square Bottle with Handle | 5 L | 6 Bottles |
£433.00
Pack of 6 |
In Stock
Log in to see stock. |
|||
Description
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
- Durable, crystal clear PETG for sample integrity
- Closures with polyethylene foam core liners for leakproof† assurance
- Custom options available for fluid transfer applications
- Supported by extensive validation documentation
- Suitable for storage in conditions from -40° to +70°C
Warranty: 90 days
Specifications
Clear | |
PETG | |
-70°C to 70°C | |
Yes | |
Yes | |
Square |
Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation | |
Sterile | |
USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315 | |
Not Autoclavable | |
Narrow |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.