Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
Brand: Thermo Scientific™ 342032-0010
416.11 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 55
Additional Details : Weight : 9.70000kg
Description
Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).
Features:
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
- Leakproof† with Nalgene lined continuous-thread closures (Cat. No. 342158)
Ordering Information:
Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).
Specifications
Serum Vials | |
Clear | |
PETG | |
10 mL | |
252 | |
Nalgene | |
Lined Continuous-thread Closure (Sold Separately) | |
1260 Cases, 252 Pk. | |
Serum Vial | |
Lined Continuous-Thread Closure |
11 mm | |
24 mm | |
Sterile | |
1260 | |
1/3 oz. | |
10 mL | |
54 mm | |
ISO 13485:2016 | |
20-415 |