missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays

Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:
- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile
10-6
SAL - Molded in graduations
- Closure supplied separately
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 8 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | Clear |
Description | Media bottles without closure |
Diameter (English) | 2.3 in. |
Diameter (English) Inner Neck | 1.1 in. |
Diameter (Metric) Inner Neck | 28 mm |
For Use With (Application) | Filling Lines |
Material | PETG |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1323575 | Certificate of Compliance | 10/12/2024 | 342024-0250 |
1357191 | Certificate of Processing | 04/12/2024 | 342024-0250 |
1375524 | Certificate of Processing | 04/12/2024 | 342024-0250 |
1323575 | Certificate of Processing | 04/12/2024 | 342024-0250 |
1378485 | Certificate of Processing | 04/12/2024 | 342024-0250 |
Special Offers
-
Multiple Manufacturers
Outlet Corner
Use code 24095 to receive this promotion.Promo Code: 24095Expires: 2025-04-15
Days
15Hours
15Minutes
47Seconds
51
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays >
Spot an opportunity for improvement?Share a Content Correction