missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure Product Code.: 10420613
In Stock
to see stock.
Add to basket

Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
GREENER_CHOICE

Product Code. 10420613
250 mL
Click to view available options
Nominal Capacity:
250 mL
450 mL
This item is not returnable. View return policy

Product Code. 10420613

Brand: Thermo Scientific™ 31400250

In Stock
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.

Ideal for large-volume processes such as cell harvesting, pelleting, and protein purification, Thermo Scientific™ Nalgene™ PC Centrifuge Bottles are molded of crystal-clear polycarbonate, which allows easy sample visualization and results in excellent mechanical strength. These centrifuge bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors.

  • Molded of polycarbonate for easy visualization of samples, these bottles are ideal for low- to moderate-speed centrifugation of biological and chemical samples
  • Polypropylene sealing cap closure with silicone gasket adds leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity

Ordering Information:
When spinning 250mL bottle in a Thermo Scientific™ Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Material Polycarbonate
No. per Case 36
No. per Pack 4
Nominal Capacity 250 mL
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Max. Force 27,500 x g
Shape Round
Height (Metric) 134.4 mm
Product Type Centrifuge Bottle
Width (English) 2.432 in.
Length (English) 2.432 in.
Height (English) 5.29 in.
Closure Material Polypropylene, red rubber
Diameter (Metric) 61.5 mm
Closure Type Sealing Cap w/Silicone Gasket
Through the Closure Dispensing No
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1411669 Certificate TypeCertificate of Compliance Date30/08/2024 Catalog Number3140-0250
Lot Number1409688 Certificate TypeCertificate of Compliance Date31/07/2024 Catalog Number3140-0250
Lot Number1398917 Certificate TypeCertificate of Compliance Date05/01/2024 Catalog Number3140-0250
Lot Number1396001 Certificate TypeCertificate of Compliance Date30/12/2023 Catalog Number3140-0250
Lot Number1384396 Certificate TypeCertificate of Compliance Date17/05/2023 Catalog Number3140-0250

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.