Learn More
Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels
Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.
Brand: Thermo Scientific™ 6319-0040
248.64 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.
Features:
- Durable polyolefin labels
- Rated to −29°C
- Resistant to ultraviolet light, mildew, chemicals and dirt, and won't discolor, crack or tear
- Excellent for labeling microscopic slides—adhesive backing sticks well to any plastic, glass or metal surface
- Chemically compatible with most lab reagents, although solvents may dissolve adhesive
- Autoclavable
- Formatted for easy setup in word processing programs; instructions included in each pack
Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.
Specifications
Polyolefin Label, 2 x 4 | |
Polyolefin | |
500/Cs. | |
4 in. | |
2 in. |
Label | |
10 | |
100/Pk. | |
102 mm | |
51 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.