missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Brand: Thermo Scientific™ 4252-0080
97.29 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
- Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
- Suitable for general lab use, it is a durable replacement for glass funnels
- Autoclavable
Specifications
79mm | |
18 mm | |
85 mm | |
Funnel Powder PP 80mm | |
12/Pk.; 36/Cs. |
115 mL | |
Polypropylene | |
Autoclavable | |
30 mm |