Learn More
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Brand: Thermo Scientific™ 4252-0100
75.45 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 0.90000kg
Description
Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
- Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
- Suitable for general lab use, it is a durable replacement for glass funnels
- Autoclavable
Specifications
100 mm | |
23 mm | |
105 mm | |
Funnel Powder PP 100mm | |
6/Pk.; 24/Cs. |
243 mL | |
Polypropylene | |
Yes | |
34 mm |