missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles Product Code.: 10390761
In Stock
to see stock.
Add to basket

Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles
GREENER_CHOICE

Product Code. 10390761
125 mL, 28 mm
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
28 mm
38 mm
48 mm
53 mm
63 mm
This item is not returnable. View return policy

Product Code. 10390761

Brand: Thermo Scientific™ 20090004

In Stock
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Protect light-sensitive liquids or dry materials from exposure with these space-efficient, amber HDPE bottles, featuring sturdy walls and a wide mouth for easy filling.

Protect light-sensitive liquids or dry materials from exposure. Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles take up less shelf space than traditional round bottles. These high-density, polyethylene bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Excellent chemical resistance to most acids, bases and alcohols makes this container great for sampling, shipping or storing liquids or dry materials that are sensitive to UV light.

  • Amber polypropylene screw closure
  • Amber HDPE reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 light transmission requirements
  • Rectangular shape is great for space-saving use in the freezer to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Leakproof†
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Spécification

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Amber
Description Amber Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck 0.8 in.
Diameter (Metric) Inner Neck 21 mm
Material HDPE
Length (Metric) 61 mm
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Width (Metric) 38 mm
Closure Color Opaque Amber
Certifications/Compliance Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure Included Yes
Graduated No
Mouth Wide
Shape Rectangular
Height (Metric) 102 mm
Product Type Wide Mouth Bottle
Height (English) 4 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 61 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Afficher plus Afficher moins

Manuals

Package Inserts

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1416424 Certificate TypeCertificate of Compliance Date05/11/2024 Catalog Number2009-0004
Lot Number1411282 Certificate TypeCertificate of Compliance Date10/10/2024 Catalog Number2009-0004
Lot Number1399722 Certificate TypeCertificate of Compliance Date25/01/2024 Catalog Number2009-0004
Lot Number1389948 Certificate TypeCertificate of Compliance Date27/07/2023 Catalog Number2009-0004
Lot Number1373266 Certificate TypeCertificate of Compliance Date13/12/2022 Catalog Number2009-0004

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis