Learn More
Thermo Scientific™ Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Process tubes at their highest rated speed level (10,000 x g or greater) without leakage, even when full with these sealing cap assemblies.
Brand: Thermo Scientific™ DS3131-0024
33.66 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 55
Additional Details : Weight : 0.03600kg
Description
Process Thermo Scientific™ Nalgene™ FEP Oakridge 30mL and 50mL centrifuge tubes at their highest rated speed level (10,000 x g and over) without leakage, even when full. Thermo Scientific™ Nalgene™ Sealing Cap Assemblies include a Viton™ fluoroelastomer o-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Features:
- Top fits securely for use at maximum speeds without leakage
- Autoclavable/biohazard/leakproof†
Includes:
- Polypropylene screw closure
- Tefzel™ ETFE plug
- Viton fluoroelastomer o-ring
Specifications
Centrifuge Tube Cap | |
Autoclavable | |
Polypropylene, ETFE, Fluoroelastomer | |
2 | |
Closure size 24 |
24 mm | |
Sealing cap assembly | |
2 | |
Nalgene™ |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.