Learn More
Thermo Scientific™ Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Process tubes at their highest rated speed level (10,000 x g or greater) without leakage, even when full with these sealing cap assemblies.
£63.16 - £65.00
Specifications
Product Type | Centrifuge Tube Cap |
---|---|
Autoclavable | Autoclavable |
Description | Sealing cap assembly |
Material | Polypropylene, ETFE, Fluoroelastomer |
No. per Pack | 2 |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10602641
|
Thermo Scientific™
DS3131-0020 |
Closure size 20 |
£65.00
Pack of 2 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
10383341
|
Thermo Scientific™
DS3131-0024 |
Closure size 24 |
£63.16
Pack of 2 |
In Stock
Log in to see stock. |
|||||
Description
Process Thermo Scientific™ Nalgene™ FEP Oakridge 30mL and 50mL centrifuge tubes at their highest rated speed level (10,000 x g and over) without leakage, even when full. Thermo Scientific™ Nalgene™ Sealing Cap Assemblies include a Viton™ fluoroelastomer o-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Features:- Top fits securely for use at maximum speeds without leakage
- Autoclavable/Biohazard/Leakproof† when used with Nalgene FEP Oakridge tubes
Includes:
- Polypropylene screw closure
- Tefzel™ ETFE plug
- Viton™ fluoroelastomer o-ring
Specifications
Centrifuge Tube Cap | |
Sealing cap assembly | |
2 | |
Nalgene™ |
Autoclavable | |
Polypropylene, ETFE, Fluoroelastomer | |
2 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.