Learn More
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle
Thermo Scientific™ Nalgene™ Square PETG Media Bottles are certified sterile, non-pyrogenic, and ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
£643.00 - £1650.00
Specifications
Color Clear
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Material PETG
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
View More Specs
Products 7
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16480601
View Documents
Thermo Scientific™
CE-N2019-0030
30 mL
£643.00
Pack of 70
In Stock
to see stock.
Add to basket
16490601
View Documents
Thermo Scientific™
CE-N2019-0060
60 mL
£828.00
Pack of 72
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
16400611
View Documents
Thermo Scientific™
CE-N2019-0125
125 mL
£772.00
Pack of 50
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
16410611
View Documents
Thermo Scientific™
CE-N2019-0250
250 mL
£1225.00
Pack of 48
In Stock
to see stock.
Edge
Add to basket
16420611
View Documents
Thermo Scientific™
CE-N2019-0500
500 mL
£689.00
Pack of 70
In Stock
to see stock.
Add to basket
16430611
View Documents
Thermo Scientific™
CE-N2019-1000
1 L
£1650.00
Pack of 35
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
16440611
View Documents
Thermo Scientific™
CE-N2019-2000
2 L
£1635.00
Pack of 20
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
Description
Description
Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
  • Eliminates costly washing, depyrogenation and autoclaving steps
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
  • Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
  • Supported by extensive validation documentation
Specifications
Specifications
Clear
PETG
-70°C to 70°C
USP Class VI Compliant, Certified Radiation Stabilized (SAL 10-6)
Narrow
Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Sterile
<1/3 USP <788> (Ultralow particulate)
Not Autoclavable
Media Bottle
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle