missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Brand:  Thermo Scientific™ 176-0045

132.66 GBP valid until 2024-12-31
Use promo code "21934" to get your promotional price.



Code : 54

Additional Details : Weight : 0.06392kg

 View more versions of this product

Product Code. 10722732

  • £171.02 / Pack of 100
Estimated Shipment: 20-11-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

Specifications

Specifications

Green
Nylon
Non-sterile
Polypropylene
Syringe Filter
4 mm
Green; Membrane: nylon; 0.45um pore
0.45 μm
Sterilizing cryoprotectants such as DMSO and with
Nalgene™
10 to 100 mL (Sample)
4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers