Learn More
Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
£171.02 - £410.00
Product Code | Brand | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity & Availability | ||
15362378
|
Thermo Scientific™
721-1320 |
13 mm | PTFE | 0.2 μm | Non-sterile |
£259.01
Pack of 100 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
15347006
|
Thermo Scientific™
721-1345 |
13 mm | PTFE | 0.45 μm | Non-sterile |
£410.00
Pack of 100 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
15372378
|
Thermo Scientific™
722-2000 |
25 mm | Glass Fiber | 1.1 μm | Non-sterile |
£174.00
Pack of 50 |
In Stock
Log in to see stock. |
||
15352388
|
Thermo Scientific™
724-2020 |
25 mm | SFCA | 0.2 μm | Non-sterile |
£392.16
Pack of 100 |
In Stock
Log in to see stock. |
||
15362388
|
Thermo Scientific™
724-2045 |
25 mm | SFCA | 0.45 μm | Non-sterile |
£380.00
Pack of 100 |
In Stock
Log in to see stock. |
||
15382388
|
Thermo Scientific™
724-2080 |
25 mm | Cellulose Acetate | 0.8 μm | Non-sterile |
£380.00
Pack of 100 |
In Stock
Log in to see stock. |
||
15342398
|
Thermo Scientific™
727-2020 |
25 mm | Nylon | 0.2 μm | Non-sterile |
£305.99
Pack of 100 |
In Stock
Log in to see stock. |
||
15352398
|
Thermo Scientific™
727-2045 |
25 mm | Nylon | 0.45 μm | Non-sterile |
£305.99
Pack of 100 |
In Stock
Log in to see stock. |
||
15362398
|
Thermo Scientific™
728-2020 |
25 mm | PTFE | 0.2 μm | Non-sterile |
£346.00
Pack of 100 |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||
15372398
|
Thermo Scientific™
728-2045 |
25 mm | PTFE | 0.45 μm | Non-sterile |
£335.42
Pack of 100 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
10353291
|
Thermo Scientific™
176-0020 |
4 mm | Nylon | 0.2 μm | Non-sterile |
£319.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10722732
|
Thermo Scientific™
176-0045 |
4 mm | Nylon | 0.45 μm | Non-sterile |
£171.02
Pack of 100 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
Cellulose Acetate (CA)
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
Nylon
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Non-sterile | |
Syringe Filter |