missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Unwire™ Polypropylene Test Tube Racks
GREENER_CHOICE

Hold a full load of test tubes or centrifuge tubes with these polypropylene test tube racks, molded in one piece.

Brand:  Thermo Scientific™ 5976-0016

26.76 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10470051

  • £40.15 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Hold a full load of test tubes or centrifuge tubes with Thermo Scientific™ Nalgene™ Unwire™ Polypropylene Test Tube Racks. Racks are molded in one piece to provide extra strength and durability for room temperature applications.

Features:
  • Higher specific gravity than water so rack stays submerged in a water bath
  • Holds a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm sizes or up to 72 smaller diameter tubes
  • Available in four bright colors to help keep samples organized
  • Accepts large labels for easy sample identification

Note:

These racks are not autoclavable.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Test Tube Rack
6 x 12
White
Polypropylene
Rectangular
16mm Tubes
24.9 cm
12.7 cm
6.7 cm
16 mm
Test Tube Rack Unwire™ Wht PP 16mm
Not Autoclavable
Tubes (16 mm)
72
3 Tiers
9.75 x 5 x 2.75 in. (24.9 x 12.7 x 6.7cm)
9.75 in.
5 in.
2.75 in.
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Unwire™ Polypropylene Test Tube Racks > Unwire Test Tube Racks, polypropylene, for 16mm Tubes; Holds 72; White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.