Learn More
Thermo Scientific™ Nalgene™ Vacuum Chamber
Eliminate the risk of implosion or shattering at full vacuum with these vacuum chambers. Useful as a controlled environment for degassing operations and freeze drying, vacuum chambers are available with transparent PC or amber PEI bell jar.
Brand: Thermo Scientific™ 5305-0609
243.90 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Alert:
Do not expose jar to partially halogenated hydrocarbons or their vapors. Avoid direct exposure of all chemicals to the polycarbonate plate. Do not autoclave when used for vacuum service. Do not use the polycarbonate jar (5305-0609) with organic solvents, acids or bases.
Description
Eliminate the risk of implosion or shattering at full vacuum with Thermo Scientific™ Nalgene™ Vacuum Chambers. Useful as a controlled environment for degassing operations and freeze drying, vacuum chambers are available with transparent PC or amber PEI bell jar.
Features:- Useful as a controlled environment chamber for degassing operations and freeze drying
- Available as polycarbonate or polyetherimide with white polypropylene or polycarbonate plate
- Transparent
Includes:
- Transparent PC or Amber PEI bell jar
- Gasketed white polycarbonate vacuum plate
Specifications
Nalgene™ Vacuum Chambers | |
Polypropylene | |
1 | |
211 mm | |
4.7 L | |
6.63 in. | |
9.38 in. | |
Polycarbonate |
Transparent PC bell jar; Gasketed white polycarbonate vacuum plate | |
2 | |
178 mm | |
1.25 gal. | |
Transparent Clear | |
170 mm | |
237 mm | |
Vacuum Chamber |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.