missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Vacuum Chamber
GREENER_CHOICE

Eliminate the risk of implosion or shattering at full vacuum with these vacuum chambers. Useful as a controlled environment for degassing operations and freeze drying, vacuum chambers are available with transparent PC or amber PEI bell jar.

Brand:  Thermo Scientific™ 5305-0910

271.07 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10626181

  • £486.00 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Do not expose jar to partially halogenated hydrocarbons or their vapors. Avoid direct exposure of all chemicals to the polycarbonate plate. Do not autoclave when used for vacuum service. Do not use the polycarbonate jar (5305-0609) with organic solvents, acids or bases.

This item is not returnable. View return policy

Description

Description

Eliminate the risk of implosion or shattering at full vacuum with Thermo Scientific™ Nalgene™ Vacuum Chambers. Useful as a controlled environment for degassing operations and freeze drying, vacuum chambers are available with transparent PC or amber PEI bell jar.

Features:
  • Useful as a controlled environment chamber for degassing operations and freeze drying
  • Available as polycarbonate or polyetherimide with white polypropylene or polycarbonate plate
  • Transparent

Includes:

  • Transparent PC or Amber PEI bell jar
  • Gasketed white polycarbonate vacuum plate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nalgene™ Vacuum Chambers
Polycarbonate
1
330 mm
8.3 L
8.75 in.
10 in.
Polyetherimide
Amber PEI bell jar; Gasketed white polycarbonate vacuum plate
1
321 mm
2.625 gal.
Transparent Amber
222 mm
254 mm
Vacuum Chamber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.